Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MED19 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP17959720UL
Description
MED19 Polyclonal specifically detects MED19 in Human, Rat samples. It is validated for Western Blot.Specifications
MED19 | |
Polyclonal | |
Western Blot 1:1000 | |
D3ZAP1 | |
MED19 | |
Synthetic peptide corresponding to a region of Rat MED19 (NP_001101211). Peptide sequence MRELPGSTELTGSTNLITHYNLEQAYNKFCGKKVKEKLSNFLPDLPGMID. | |
Affinity Purified | |
RUO | |
219541 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
LCMR1mediator of RNA polymerase II transcription subunit 19, Lung cancer metastasis-related protein 1, mediator complex subunit 19DT2P1G7, mediator of RNA polymerase II transcription, subunit 19 homolog, mediator of RNA polymerase II transcription, subunit 19 homolog (S. cerevisiae) | |
Rabbit | |
27 kDa | |
20 μL | |
Primary | |
Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction