Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MED19 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $487.50
Specifications
Antigen | MED19 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17959720
![]() |
Novus Biologicals
NBP17959720UL |
20 μL |
Each for $158.00
|
|
|||||
NBP179597
![]() |
Novus Biologicals
NBP179597 |
100 μL |
Each for $487.50
|
|
|||||
Description
MED19 Polyclonal specifically detects MED19 in Human, Rat samples. It is validated for Western Blot.Specifications
MED19 | |
Polyclonal | |
Rabbit | |
D3ZAP1 | |
219541 | |
Synthetic peptide corresponding to a region of Rat MED19 (NP_001101211). Peptide sequence MRELPGSTELTGSTNLITHYNLEQAYNKFCGKKVKEKLSNFLPDLPGMID. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
LCMR1mediator of RNA polymerase II transcription subunit 19, Lung cancer metastasis-related protein 1, mediator complex subunit 19DT2P1G7, mediator of RNA polymerase II transcription, subunit 19 homolog, mediator of RNA polymerase II transcription, subunit 19 homolog (S. cerevisiae) | |
MED19 | |
IgG | |
27 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title