Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MED19 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | MED19 |
---|---|
Applications | Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
MED19 Polyclonal specifically detects MED19 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
MED19 | |
Polyclonal | |
Rabbit | |
Human | |
LCMR1mediator of RNA polymerase II transcription subunit 19, Lung cancer metastasis-related protein 1, mediator complex subunit 19DT2P1G7, mediator of RNA polymerase II transcription, subunit 19 homolog, mediator of RNA polymerase II transcription, subunit 19 homolog (S. cerevisiae) | |
MED19 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunofluorescence, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
219541 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:GSHDNSSLRSLIEKPPILSSSFNPITGTMLAGFRLHTGPLPEQCRLMHIQPPKKKNKHKHKQSRTQDPV | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title