Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MED19 Rabbit anti-Human, Janelia Fluor 549, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB044180 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB044180 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB044180 Supplier Novus Biologicals Supplier No. NBP297489JF549

Rabbit Polyclonal Antibody

MED19 Polyclonal specifically detects MED19 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen MED19
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias LCMR1mediator of RNA polymerase II transcription subunit 19, Lung cancer metastasis-related protein 1, mediator complex subunit 19DT2P1G7, mediator of RNA polymerase II transcription, subunit 19 homolog, mediator of RNA polymerase II transcription, subunit 19 homolog (S. cerevisiae)
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MED19 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 219541
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.