Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MED21 Antibody (PCRP-MED21-4B5), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB178980 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB178980 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB178980 Supplier Novus Biologicals Supplier No. NBP314069AF750
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MED21 Monoclonal antibody specifically detects MED21 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen MED21
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-MED21-4B5
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias hSrb7, mediator complex subunit 21SRB7 (suppressor of RNA polymerase B, yeast) homolog, RNA polymerase II complex component SRB7, RNA polymerase II holoenzyme component SRB7, RNAPII complex component SRB7, SRB7 suppressor of RNA polymerase B homolog, SRB7SRB7 suppressor of RNA polymerase B homolog (yeast), SURB7mediator of RNA polymerase II transcription subunit 21
Host Species Mouse
Immunogen Recombinant full-length human MED21 protein (Uniprot: Q13503)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 9412
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.