Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MED28 Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB044249 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB044249 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB044249 Supplier Novus Biologicals Supplier No. NBP297494V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MED28 Polyclonal specifically detects MED28 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen MED28
Applications Immunofluorescence
Classification Polyclonal
Conjugate DyLight 405
Dilution Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias DKFZP434N185, EG1DKFZp434N185, magicin, mediator complex subunit 281500003D12Rik, mediator of RNA polymerase II transcription, subunit 28 homolog (S. cerevisiae), subunit 28 homolog
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human MED28 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80306
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.