Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MED28 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB357019 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB357019 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB357019 Supplier Novus Biologicals Supplier No. NBP176931PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MED28 Polyclonal antibody specifically detects MED28 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen MED28
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias DKFZP434N185, EG1DKFZp434N185, magicin, mediator complex subunit 281500003D12Rik, mediator of RNA polymerase II transcription, subunit 28 homolog (S. cerevisiae), subunit 28 homolog
Host Species Rabbit
Immunogen Antibody was raised against a 17 amino acid synthetic peptide from near the center of human MED28. The immunogen is located within amino acids 50 - 100 of MED28. Amino Acid Squence: SQDYVNGTDQEEIRTGVDQ
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80306
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.