Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MED30 Antibody (OTI3B3), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N272619A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272619A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272619A700 Supplier Novus Biologicals Supplier No. NBP272619AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MED30 Monoclonal specifically detects MED30 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen MED30
Applications Western Blot
Classification Monoclonal
Clone OTI3B3
Conjugate Alexa Fluor 700
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias mediator complex subunit 30TRAP/Mediator complex component TRAP25, mediator of RNA polymerase II transcription subunit 30, putative mediator of RNA polymerase II transcription subunit 30, THRAP6MGC9890, thyroid hormone receptor associated protein 6, Thyroid hormone receptor-associated protein 6, Thyroid hormone receptor-associated protein complex 25 kDa component, Trap25, TRAP25MED30S
Gene Symbols MED30
Host Species Mouse
Immunogen Full length recombinant protein of human MED30 (NP_542382) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 90390.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.