Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MED30 Antibody (OTI3B3), PE, Novus Biologicals™
SDP

Catalog No. NB287935 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287935 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287935 Supplier Novus Biologicals Supplier No. NBP272619PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MED30 Monoclonal antibody specifically detects MED30 in Human, Mouse, Rat samples. It is validated for Western Blot

Specifications

Antigen MED30
Applications Western Blot
Classification Monoclonal
Clone OTI3B3
Conjugate PE
Dilution Western Blot
Formulation PBS
Gene Alias mediator complex subunit 30TRAP/Mediator complex component TRAP25, mediator of RNA polymerase II transcription subunit 30, putative mediator of RNA polymerase II transcription subunit 30, THRAP6MGC9890, thyroid hormone receptor associated protein 6, Thyroid hormone receptor-associated protein 6, Thyroid hormone receptor-associated protein complex 25 kDa component, Trap25, TRAP25MED30S
Host Species Mouse
Immunogen Full length human recombinant protein of human MED30 (NP_542382) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 90390
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.