Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MED7 Antibody (PCRP-MED7-1B8), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB179021 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB179021 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB179021 Supplier Novus Biologicals Supplier No. NBP314071AF350
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

MED7 Monoclonal antibody specifically detects MED7 in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence

Specifications

Antigen MED7
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Monoclonal
Clone PCRP-MED7-1B8
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Gene Alias Activator-recruited cofactor 34 kDa component, Cofactor required for Sp1 transcriptional activation subunit 9, cofactor required for Sp1 transcriptional activation, subunit 9 (33kD), cofactor required for Sp1 transcriptional activation, subunit 9, 33kDa, CRSP33CRSP complex subunit 9, CRSP9hMED7, mediator complex subunit 7ARC34, mediator of RNA polymerase II transcription subunit 7, MGC12284, RNA polymerase transcriptional regulation mediator subunit 7 homolog, Transcriptional coactivator CRSP33
Host Species Mouse
Immunogen Recombinant full-length human MED7 protein (Uniprot: O43513)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline DNA Replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 9443
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.