Learn More
Stoelting™ Medbond Tissue Glue
SDP
Stoelting™ Medbond is a hemostatic agents that seals off capillary ends.
$259.00 - $1250.00
Specifications
For Use With (Application) Neuroscience
Size 2 mL
View More Specs
Products 2
Catalog Number Mfr. No. Includes Price Quantity & Availability  
Catalog Number Mfr. No. Includes Price Quantity & Availability  
10-000-647
SDP
View Documents
Stoelting™
50478
Pack of 5
Pack of 5 for $259.00
Only null left
Add to Cart
10-000-648
SDP
View Documents
Stoelting™
50479
Box of 25
Pack of 25 for $1,250.00
Only null left
Add to Cart
Description
Description
  • Monomeric formulation which upon contact with an alkaline pH environment, polymerizes to form a thin flexible waterproof bandage.
  • Polymerizes in apporximately one second after contact with moist wound surfaces.
  • Polymerization is slower on a dry field.
  • Also seals off exposed nerve endings.
Specifications
Specifications
Neuroscience
2 mL
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.