Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Medi-First™ Extra-Strength Pain-Relief Tablets

Catalog No. p-7078220 Shop All Medique Products
Click to view available options
Quantity:
2/Pk., 125/Box
2/Pk., 250/Box

Catalog No. 19-090-803

Supplier: Medique 81148

Relieves symptoms associated with headaches, muscular aches, pain, fever and menstrual cramps.

  • Each tablet contains Acetaminophen 110mg, Salicylamide 152mg, Caffeine 32.4mg

Specifications

Product Type Extra Strength Pain Reliever
Active Ingredients Acetaminophen (110 mg), Salicylamide (152 mg), Caffeine (32.4 mg)
For Use With (Application) For pain, headache, cramp and fever relief
Quantity 2/Pk., 125/Box
Recommended Age 12 years and older
Storage Requirements 59°F to 86°F (15°C to 30°C)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.