Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medique Medi-First™ Instant Cold Packs

Catalog No. p-7078242 Shop All Medique Products
Click to view available options
Size:
Small
Large
Dimensions (L x W):
4 x 6 in.
5 x 9 in.
Format:
Boxed
Unboxed
Unitized Box

For fast pain relief.

  • Instant cold compress

Specifications

For Use With (Application) For fast pain relief
Product Type Instant Cold Pack
Format Boxed
Size Large
Dimensions (L x W) 5 x 9 in.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.