Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Medique Medi-First™ Metal Detectable Blue Bandages
Shop All Medique Products
Click to view available options
Style:
Plastic Strip
Woven Strip
Description
- Designed to be highly visible
- Latex-free
Specifications
Specifications
For Use With (Application) | food and restaurant preparations |
Product Type | Metal-detectable Bandage |
Color | Blue |
Latex Free | Yes |
Quantity | 100 Pk. |
Style | Plastic Strip |
Dimensions (L x W) | 1 x 3 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction