Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Medique Medi-First™ Water-Proof First-Aid Tape
Shop All Medique Products
Click to view available options
Dimensions (L x W):
10 yd. x 0.5 in.
10 yd. x 1 in.
5 yd. x 0.5 in.
5 yd. x 1 in.
Description
- With adhesive
- Adheres securely and tears easily
- Latex-free
Specifications
Specifications
| For Use With (Application) | Holds gauze and dressings in place |
| Product Type | First Aid Tape |
| Adhesive | Yes |
| Latex Free | Yes |
| Waterproof | Yes |
| Dimensions (L x W) | 10 yd. x 1 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction