Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

mediator of cell motility 1 Antibody (1E9), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP22293A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP22293A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP22293A30 Supplier Novus Biologicals Supplier No. NBP222593AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

mediator of cell motility 1 Monoclonal specifically detects mediator of cell motility 1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.

Specifications

Antigen mediator of cell motility 1
Applications Western Blot, Flow Cytometry, ELISA
Classification Monoclonal
Clone 1000000000
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, ELISA
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias C21orf19-like protein, C2orf4DKFZp434I0135, CGI-27, FLJ25031, HCV NS5A-transactivated protein 7, Hepatitis C virus NS5A-transactivated protein 7, mediator of cell motility 1, Mediator of ErbB2-driven cell motility 1, memo-1, MEMOchromosome 2 open reading frame 4, NS5ATP7, protein MEMO1
Gene Symbols MEMO1
Host Species Mouse
Immunogen Recombinant human MEMO1 (1-297aa) purified from E. coli
Quantity 0.1 mL
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 51072
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.