Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medroxyprogesterone acetate, 97%, Thermo Scientific Chemicals

Catalog No. ac461120010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461120010 1 g
AC461120050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC461120010 Supplier Thermo Scientific Chemicals Supplier No. 461120010
Only null left
Add to Cart
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-58-9
Molecular Formula C24H34O4
Molecular Weight (g/mol) 386.52
InChI Key PSGAAPLEWMOORI-PEINSRQWSA-N
Synonym medroxyprogesterone acetate, medroxyprogesterone 17-acetate, metigestrona, provera, depo-provera, gestapuran, farlutin, perlutex, veramix, methylacetoxyprogesterone
PubChem CID 6279
ChEBI CHEBI:6716
IUPAC Name [(6S,8R,9S,10R,13S,14S,17R)-17-acetyl-6,10,13-trimethyl-3-oxo-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthren-17-yl] acetate
SMILES CC1CC2C(CCC3(C2CCC3(C(=O)C)OC(=O)C)C)C4(C1=CC(=O)CC4)C

Specifications

Melting Point 206.0°C to 208.0°C
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble.
Formula Weight 386.52
Percent Purity 97%
Chemical Name or Material Medroxyprogesterone acetate
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Suspected of causing cancer.
May damage fertility.
May damage the unborn child.
Precautionary Statement GHS P Statement Obtain special instructions before use. Wear protective gloves/protective clothing/eye protection/face protection. IF exposed or concerned: Get medical advice/attention.
EINECSNumber 200-757-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.