Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEF2C, p Ser222 Antibody, Novus Biologicals™
SDP

Catalog No. NBP260774 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP260774 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP260774 Supplier Novus Biologicals Supplier No. NBP260774
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MEF2C Polyclonal specifically detects MEF2C in Human, Mouse, Rat, Bovine, Chicken, Guinea Pig, Primate, Sheep samples. It is validated for Western Blot.

Specifications

Antigen MEF2C (p Ser222)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation 10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per ml BSA and 50% glycerol. with No Preservative
Gene Alias DEL5q14.3, myocyte enhancer factor 2C, myocyte-specific enhancer factor 2C, polypeptide C
Gene Symbols MEF2C
Host Species Rabbit
Immunogen Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser222 conjugated to KLH. Accession # Q06413
Molecular Weight of Antigen 51 kDa
Purification Method Antigen Affinity-purified
Quantity 100 μL
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4208
Target Species Human, Mouse, Rat, Bovine, Chicken, Guinea Pig, Primate, Sheep
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.