Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEFV Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NB163478 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB163478 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB163478 Supplier Novus Biologicals Supplier No. NBP280113R
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MEFV Polyclonal antibody specifically detects MEFV in Human samples. It is validated for Western Blot

Specifications

Antigen MEFV
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias FMF, FMFmarenostrin, Marenostrin, Mediterranean fever, MEFpyrin, MGC126560, MGC126586, TRIM20
Host Species Rabbit
Immunogen Synthetic peptide corresponding to aa 2-29 (A2KTPSDHLLSTLEELVPYDFEKFKFKLQ29) of human MEFV.
Purification Method Epitope affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4210
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.