Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEFV Antibody, Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB163477 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB163477 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB163477 Supplier Novus Biologicals Supplier No. NBP280113JF646

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

MEFV Polyclonal antibody specifically detects MEFV in Human samples. It is validated for Western Blot

Specifications

Antigen MEFV
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias FMF, FMFmarenostrin, Marenostrin, Mediterranean fever, MEFpyrin, MGC126560, MGC126586, TRIM20
Host Species Rabbit
Immunogen Synthetic peptide corresponding to aa 2-29 (A2KTPSDHLLSTLEELVPYDFEKFKFKLQ29) of human MEFV.
Purification Method Epitope affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology, Cytoskeleton Markers, Immunology, Innate Immunity, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4210
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.