Learn More
MEI1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 134561AP150UL
Description
This antibody recognizes isoform 1, 2, 3, 5, and 7 of MEI1 gene with MW 141 kDa, 67-72 kDa, 49-54 kDa, and 26 kDa.
The predomit cause of spermatogenic arrest of meiosis is the failure of homologous chromosomes to accurately synapse. MEI1 (Meiosis inhibitor protein 1), also designated Meiosis defective protein 1, is a 1274 amino acid protein that is likely required for the formation of genetically programmed double-strand breaks, the first step in the initiation of meiosis. With predomit expression in testis, it is likely that defects of the gene encoding MEI1 results in male infertility. Interestingly, studies show that genetic variation in the MEI gene possibly predisposes European Americans but not Israeli men to infertility by meiotic arrest. Human MEI1 shares 79% sequence similarity with its mouse homolog. There are seven isoforms of MEI1 that are produced as a result of alternative splicing events.Specifications
| MEI1 | |
| Polyclonal | |
| Unconjugated | |
| MEI1 | |
| MEI1, Meiosis defective protein 1, meiosis inhibitor 1, Meiosis inhibitor protein 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 150365, 315162, 74369 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q5TIA1, Q9D4I2 | |
| MEI1 | |
| MEI1 Fusion Protein Ag4253 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.