Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEIG1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB164283 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB164283 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB164283 Supplier Novus Biologicals Supplier No. NBP281733AF594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MEIG1 Polyclonal antibody specifically detects MEIG1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen MEIG1
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias bA2K17.3, meiosis expressed gene 1 homolog, meiosis expressed gene 1 homolog (mouse)
Host Species Rabbit
Immunogen MEIG1 antibody was raised against a 20 amino acid synthetic peptide near the carboxy terminal of human MEIG1. The immunogen is located within the last 50 amino acids of MEIG1. Amino Acid Squence: YVKKLQRRDNTFYYYNKQRE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 644890
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.