Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Meis homeobox 3 Antibody (OTI3E12), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB272642APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB272642APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB272642APC Supplier Novus Biologicals Supplier No. NBP272642APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Meis homeobox 3 Monoclonal specifically detects Meis homeobox 3 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen Meis homeobox 3
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3E12
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias DKFZp547H236, Meis homeobox 3, Meis1, myeloid ecotropic viral integration site 1 homolog 3, Meis1, myeloid ecotropic viral integration site 1 homolog 3 (mouse), Meis1-related protein 2, MRG2homeobox protein Meis3
Gene Symbols MEIS3
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-261 of human Meis homeobox 3 (NP_064545) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 56917.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.