Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEK1/2 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB119789 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB119789 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB119789 Supplier Novus Biologicals Supplier No. NBP309166

Rabbit Polyclonal Antibody

MEK1/2 Polyclonal specifically detects MEK1/2 in Human samples. It is validated for Western Blot.

Specifications

Antigen MEK1/2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/ml
Formulation PBS
Gene Alias CFC3, CFC4, dual specificity mitogen-activated protein kinase kinase 1, dual specificity mitogen-activated protein kinase kinase 2, ERK activator kinase 1, ERK activator kinase 2, MAP kinase kinase 2, MAPK/ERK kinase 1, MAPK/ERK kinase 2, MAPKK 1, MAPKK1, MAPKK2, MEK 1, MEK1, MEK2, MEL, mitogen-activated protein kinase kinase 2, p45, MKK1, MKK2, PRKMK1, PRKMK2, protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1)
Host Species Rabbit
Immunogen Synthetic peptide (coupled with KLH) derived from aminoacid sequence 33-53 of human MAPK Kinase 1. The immunogen sequence is highly conserved within species (human, mouse, rat, hamster, bovine etc.).
Purification Method Affinity purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 5604
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.