Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEK1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP224560SS

 View more versions of this product

Catalog No. NBP224560SS


Only null left
Add to Cart

Description

Description

MEK1 Polyclonal specifically detects MEK1 in Human, Mouse, Rat, Bovine, Canine, Chicken, Drosophila, Primate samples. It is validated for Western Blot.
Specifications

Specifications

MEK1
Polyclonal
Unconjugated
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
MAP2K1
A portion of amino acids 200-250 of human MEK1 was used as the immunogen.
0.025 mg
Angiogenesis, Autophagy, Cancer, MAP Kinase Signaling, Protein Phosphatase, Signal Transduction, Tyrosine Kinases
5604
Human, Mouse, Rat, Bovine, Canine, Chicken, Drosophila, Primate
IgG
Western Blot
0.5 mg/mL
Western Blot 5 ug/ml
dual specificity mitogen-activated protein kinase kinase 1, ERK activator kinase 1, MAP kinase kinase 1, MAPK/ERK kinase 1, MAPKK1, MEK1MAPKK 1, mitogen-activated protein kinase kinase 1, MKK1MEK 1, PRKMK1EC 2.7.12.2, protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1)
Rabbit
Affinity Purified
RUO
Primary
The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, chimp, bovine, dog and sheep.
Store at -20C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only