Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEK1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.50 - $426.00
Specifications
Antigen | MEK1 |
---|---|
Concentration | 0.5 mg/mL |
Dilution | Western Blot 5 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP224560SS
![]() |
Novus Biologicals
NBP224560SS |
0.025 mg |
Each for $206.50
|
|
|||||
NBP224560
![]() |
Novus Biologicals
NBP224560 |
0.1 mg |
Each for $426.00
|
|
|||||
Description
MEK1 Polyclonal specifically detects MEK1 in Human, Mouse, Rat, Bovine, Canine, Chicken, Drosophila, Primate samples. It is validated for Western Blot.Specifications
MEK1 | |
Western Blot 5 ug/ml | |
Polyclonal | |
Rabbit | |
Angiogenesis, Autophagy, Cancer, MAP Kinase Signaling, Protein Phosphatase, Signal Transduction, Tyrosine Kinases | |
0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
5604 | |
A portion of amino acids 200-250 of human MEK1 was used as the immunogen. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
0.5 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Human, Mouse, Rat, Bovine, Canine, Chicken, Drosophila, Primate | |
dual specificity mitogen-activated protein kinase kinase 1, ERK activator kinase 1, MAP kinase kinase 1, MAPK/ERK kinase 1, MAPKK1, MEK1MAPKK 1, mitogen-activated protein kinase kinase 1, MKK1MEK 1, PRKMK1EC 2.7.12.2, protein kinase, mitogen-activated, kinase 1 (MAP kinase kinase 1) | |
MAP2K1 | |
IgG | |
Affinity Purified | |
The amino acid sequence used as immunogen is 100% homologous in human, mouse, rat, chimp, bovine, dog and sheep. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title