Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEK2 Antibody (BLR079G), Novus Biologicals™
SDP

Catalog No. NB001669 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001669 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001669 Supplier Novus Biologicals Supplier No. NBP278800
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

MEK2 Monoclonal specifically detects MEK2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunocytochemistry.

Specifications

Antigen MEK2
Applications Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Monoclonal
Clone BLR079G
Conjugate Unconjugated
Dilution Western Blot 1:1000, Immunohistochemistry 1:100-1:500, Immunoprecipitation 20 ul/mg, Immunohistochemistry-Paraffin, Immunocytochemistry 1:100-1:500
Formulation Borate Buffered Saline (BBS) pH 8.2 containing 0.1% BSA with 0.09% Sodium Azide
Gene Alias ERK activator kinase 2, MAPKK 2, MEK 2, MEK2EC 2.7.12.2, mitogen-activated protein kinase kinase 2, MKK2FLJ26075, p45, PRKMK2MAPKK2
Gene Symbols MAP2K2
Host Species Rabbit
Immunogen Recombinant Monoclonal Antibody; between 1 and 50
Purification Method Protein A or G purified from cell culture supernatant
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 5605
Target Species Human, Mouse
Content And Storage Store at 2-8°C/ 1 year from date of receipt
Form Supernatant
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.