Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEK2 Antibody (OTI8G6), Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. N272646A647 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N272646A647 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N272646A647 Supplier Novus Biologicals Supplier No. NBP272646AF647
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MEK2 Monoclonal specifically detects MEK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen MEK2
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI8G6
Conjugate Alexa Fluor 647
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ERK activator kinase 2, MAPKK 2, MEK 2, MEK2EC 2.7.12.2, mitogen-activated protein kinase kinase 2, MKK2FLJ26075, p45, PRKMK2MAPKK2
Gene Symbols MAP2K2
Host Species Mouse
Immunogen Full-length protein expressed in 293T cell transfected with human MEK2 expression vector.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Angiogenesis, Autophagy, Cancer, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Protein Phosphatase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5605.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.