Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEK2 Rabbit anti-Human, Clone: SR13-07, Novus Biologicals™
SDP

Catalog No. NBP267332 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP267332 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP267332 Supplier Novus Biologicals Supplier No. NBP267332

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Rabbit Monoclonal Antibody

MEK2 Monoclonal specifically detects MEK2 in Human, Mouse, Rat, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.

Specifications

Antigen MEK2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SR13-07
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation, Immunohistochemistry-Paraffin 1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias ERK activator kinase 2, MAPKK 2, MEK 2, MEK2EC 2.7.12.2, mitogen-activated protein kinase kinase 2, MKK2FLJ26075, p45, PRKMK2MAPKK2
Gene Symbols MAP2K2
Host Species Rabbit
Immunogen Recombinant protein within Human MEK2 aa 1-393/ 393. (SwissProt: P36507 Human; SwissProt: Q02750 Human; SwissProt: P31938 Mouse; SwissProt: Q63932 Mouse; SwissProt: P36506 Rat; SwissProt: Q01986 Rat)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Angiogenesis, Autophagy, Cancer, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Protein Phosphatase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 5605
Target Species Human, Mouse, Rat, Zebrafish
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.