Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEK5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP19747825UG
Description
MEK5 Polyclonal specifically detects MEK5 in Human, Mouse, Rat, Canine, Primate, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MEK5 | |
Polyclonal | |
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
HsT17454, MAP kinase kinase 5, MAP kinase kinase MEK5b, MAPK/ERK kinase 5, MAPKK 5, MAPKK5EC 2.7.12.2, MEK5MEK 5, mitogen-activated protein kinase kinase 5, MKK5, PRKMK5dual specificity mitogen-activated protein kinase kinase 5 | |
Rabbit | |
Affinity Purified | |
RUO | |
5607 | |
Store at -20C. Avoid freeze-thaw cycles. |
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS (pH 7.2) with 0.09% Sodium Azide | |
MAP2K5 | |
Synthetic peptide corresponding to a portion of human MEK5. | |
25 μg | |
Primary | |
Rabbit, Sheep, Human, Xenopus species, Primate, Canine, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction