Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MEK5 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$182.00 - $499.50
Specifications
Antigen | MEK5 |
---|---|
Dilution | Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 |
Applications | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB018780
![]() |
Novus Biologicals
NBP19747825UG |
25 μg |
Each for $182.00
|
|
|||||
NBP19747801
![]() |
Novus Biologicals
NBP1974780.1MG |
0.1 mg |
Each for $499.50
|
|
|||||
Description
MEK5 Polyclonal specifically detects MEK5 in Human, Mouse, Rat, Canine, Primate, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
MEK5 | |
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) with 0.09% Sodium Azide | |
5607 | |
Synthetic peptide corresponding to a portion of human MEK5. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500 | |
Polyclonal | |
Rabbit | |
Rabbit, Sheep, Human, Xenopus species, Primate, Canine, Mouse, Rat | |
HsT17454, MAP kinase kinase 5, MAP kinase kinase MEK5b, MAPK/ERK kinase 5, MAPKK 5, MAPKK5EC 2.7.12.2, MEK5MEK 5, mitogen-activated protein kinase kinase 5, MKK5, PRKMK5dual specificity mitogen-activated protein kinase kinase 5 | |
MAP2K5 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title