Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEKK4 Antibody, Novus Biologicals™
SDP

Catalog No. NBP232187 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP232187 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP232187 Supplier Novus Biologicals Supplier No. NBP232187
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

MEKK4 Polyclonal specifically detects MEKK4 in Human samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen MEKK4
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg lysate
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_005913.2
Gene Alias EC 2.7.11.25, FLJ42439, MAPKKK4yeast, homolog of, MEKK 4, mitogen-activated protein kinase kinase kinase 4
Gene Symbols MAP3K4
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 75 to 125 of human Mitogen-Activated Protein Kinase Kinase Kinase 4 using the numbering given in entry NP_005913.2 (GeneID 4216).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 4216
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.