Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melanocortin-3 R/MC3R Antibody
SDP

Catalog No. NB036615 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB036615 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB036615 Supplier Novus Biologicals Supplier No. NBP289024

Rabbit Polyclonal Antibody

Melanocortin-3 R/MC3R Polyclonal specifically detects Melanocortin-3 R/MC3R in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen Melanocortin-3 R/MC3R
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Lyophilized from PBS, pH 7.4, 1% BSA
Gene Alias BMIQ9, MC3, MC3-R, melanocortin 3 receptor, melanocortin receptor 3, OB20, Obesity quantitative trait locus, OQTL
Host Species Rabbit
Immunogen Synthetic peptide corresponding to aa 102-117 of rat Melanocortin-3 R/MC3R.
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4159
Reconstitution To lyophilized antibody, add 50 μL or 200 μL (depending on which size was purchased) of deionized water. To lyophilized peptide, add 100 μL of deionized water.
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.