Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melanocortin-3 R/MC3R Antibody (345404), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB371564 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB371564 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB371564 Supplier Novus Biologicals Supplier No. FAB3737Z
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Melanocortin-3 R/MC3R Monoclonal specifically detects Melanocortin-3 R/MC3R in Mouse samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Melanocortin-3 R/MC3R
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 345404
Conjugate DyLight 755
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias BMIQ9, MC3, MC3-R, melanocortin 3 receptor, melanocortin receptor 3, OB20, Obesity quantitative trait locus, OQTL
Host Species Rat
Immunogen Y3 rat myeloid cell line transfeced with mouse Melanocortin 3R/MC3R
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 4159
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.