Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melanocortin-5 R/MC5R Antibody (821012), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. NB310956 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310956 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310956 Supplier Novus Biologicals Supplier No. FAB8205APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Melanocortin-5 R/MC5R Monoclonal specifically detects Melanocortin-5 R/MC5R in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen Melanocortin-5 R/MC5R
Applications Flow Cytometry
Classification Monoclonal
Clone 821012
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias MC2, MC-2, MC5-R, melanocortin 5 receptor, melanocortin receptor 5, melanocortin-5 receptor
Host Species Mouse
Immunogen NS0 mouse myeloma cell line transfected with human Melanocortin-5 R/MC5R, Met1-Asp325, Accession # P33032
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 4161
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.