Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melanocortin-5 R/MC5R Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NLS1022 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NLS1022 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NLS1022 Supplier Novus Biologicals Supplier No. NLS1022
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Melanocortin-5 R/MC5R Polyclonal specifically detects Melanocortin-5 R/MC5R in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Melanocortin-5 R/MC5R
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 7.6 - 8 ug/ml
Formulation PBS with 0.1% Sodium Azide
Gene Alias MC2, MC-2, MC5-R, melanocortin 5 receptor, melanocortin receptor 5, melanocortin-5 receptor
Gene Symbols MC5R
Host Species Rabbit
Immunogen Synthetic 18 amino acid peptide from N-terminal extracellular domain of human Melanocortin-5 R/MC5R.
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 4161
Test Specificity Human Melanocortin-5 R/MC5R. BLAST analysis of the peptide immunogen showed no homology with other human proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.