Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melanoma antigen family C2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP234145 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234145 0.1 mL
NB436621 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP234145 Supplier Novus Biologicals Supplier No. NBP234145
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Melanoma antigen family C2 Polyclonal specifically detects Melanoma antigen family C2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin).

Specifications

Antigen Melanoma antigen family C2
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:500 - 1:1000
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q9UBF1
Gene Alias Cancer/testis antigen 10, CT10MAGE-E1 antigen, HCA587MAGE-C2, hepatocellular cancer antigen 587, Hepatocellular carcinoma-associated antigen 587, MAGE-C2 antigen, MAGEE1MGC13377, melanoma antigen family C, 2, melanoma antigen, family E, 1, cancer/testis specific, melanoma-associated antigen C2
Gene Symbols MAGEC2
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: PVPGVPFRNVDNDSPTSVELEDWVDAQHPTDEEEEEASSASSTLYLVFSPS
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51438
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.