Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melanoma Associated Antigen Antibody (MAA/1414), DyLight 350, Novus Biologicals™
SDP

Catalog No. NB390083 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB390083 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB390083 Supplier Novus Biologicals Supplier No. NBP308848UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Melanoma Associated Antigen Monoclonal antibody specifically detects Melanoma Associated Antigen in Human samples. It is validated for ELISA

Specifications

Antigen Melanoma Associated Antigen
Applications ELISA
Classification Monoclonal
Clone MAA/1414
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias Chronic μgcerative stomatitis protein, CUSP, Keratinocyte transcription factor KET, p40, p51, p63, p73L, TP63, Transformation-related protein 63, Tumor protein 63, Tumor protein p73-like
Host Species Mouse
Immunogen proteins extracted from a melanoma
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.