Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melanoma Marker (MART-1 + gp100) Antibody (DT101+ BC199 + HMB45), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP234608IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234608IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP234608IR Supplier Novus Biologicals Supplier No. NBP234608IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Melanoma Marker (MART-1 + gp100) Monoclonal specifically detects Melanoma Marker (MART-1 + gp100) in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Melanoma Marker (MART-1 + gp100)
Applications Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone DT101+ BC199 + HMB45
Conjugate DyLight 755
Dilution Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Alias MART1, MART-1
Host Species Mouse
Immunogen Recombinant hMART-1 protein (DT101& BC199); Extract of pigmented melanoma metastases from lymph nodes (HMB45)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2315
Test Specificity This antibody cocktail recognizes two melanoma-specific proteins, which include MART-1 and gp100. MART-1 is a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Function of gp100 is not known but it is reported to be a useful marker for melanocytes and melanomas. A cocktail of these two markers is sensitive for labeling formalin-fixed, paraffin-embedded melanomas and other tumors showing melanocytic differentiation.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b κ, IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.