Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melanoma Marker (MART-1 + Tyrosinase + gp100) Antibody (M2-7C10 + M2-9E3 + T311 + HMB45), Novus Biologicals™
SDP

Catalog No. NBP234337B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.02 mg
0.1 mg
0.2 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234337B 0.2 mg
NBP234337F 0.02 mg
NBP234337A 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NBP234337B Supplier Novus Biologicals Supplier No. NBP2343370.2MG

Mouse Monoclonal Antibody

Melanoma Marker (MART-1 + Tyrosinase + gp100) Monoclonal antibody specifically detects Antigen in Human samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin).

Specifications

Antigen Melanoma Marker (MART-1 + Tyrosinase + gp100)
Applications Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone M2-7C10 + M2-9E3 + T311 + HMB45
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/ml
Formulation PBS with 0.05% BSA. with 0.05% Sodium Azide
Gene Accession No. Q16655
Host Species Mouse
Immunogen Recombinant hMART-1 protein (M2-7C10; M2-9E3); Recombinant tyrosinase protein (T311); Extract of pigmented melanoma metastases from lymph nodes (HMB45)
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2315
Test Specificity This antibody cocktail recognizes three melanoma-specific proteins, which include MART-1, Tyrosinase and gp100. MART-1 is a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Tyrosinase is one of the targets for cytotoxic T-cell recognition in melanoma patients. Function of gp100 is not known but it is reported to be a useful marker for melanocytes and melanomas. This cocktail of three markers is designed for extremely sensitive labeling of formalin-fixed, paraffin-embedded melanomas and other tumors showing melanocytic differentiation.
Target Species Human
Content And Storage Store at 4C.
Form Purified
Isotype IgG2b, IgG2a, IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.