Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melanophilin Antibody (OTI6E8), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB287921 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287921 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287921 Supplier Novus Biologicals Supplier No. NBP272649APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Melanophilin Monoclonal antibody specifically detects Melanophilin in Human samples. It is validated for Western Blot, Immunohistochemistry

Specifications

Antigen Melanophilin
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI6E8
Conjugate APC
Dilution Western Blot, Immunohistochemistry
Formulation PBS
Gene Alias exophilin-3, l(1)-3Rk, l1Rk3, ln, melanophilin, MGC2771, MGC59733, Slac-2a, SLAC2A, SLAC2-A, Slp homolog lacking C2 domains a, Synaptotagmin-like protein 2a
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 295-600 of human MLPH(NP_077006) produced in E.coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79083
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.