Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melanophilin Rabbit anti-Human, DyLight 405, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB104898 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB104898 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB104898 Supplier Novus Biologicals Supplier No. NBP298644V

Rabbit Polyclonal Antibody

Melanophilin Polyclonal specifically detects Melanophilin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen Melanophilin
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Formulation 50 mM Sodium Borate
Gene Alias exophilin-3, l(1)-3Rk, l1Rk3, ln, melanophilin, MGC2771, MGC59733, Slac-2a, SLAC2A, SLAC2-A, Slp homolog lacking C2 domains a, Synaptotagmin-like protein 2a
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Melanophilin fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79083
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.