Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melanotransferrin/CD228/MFI2 Antibody (893416), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB373490 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB373490 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB373490 Supplier Novus Biologicals Supplier No. FAB81751K
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Melanotransferrin/CD228/MFI2 Monoclonal specifically detects Melanotransferrin/CD228/MFI2 in Human samples. It is validated for CyTOF, Flow Cytometry.

Specifications

Antigen Melanotransferrin/CD228/MFI2
Applications CyTOF, Flow Cytometry
Classification Monoclonal
Clone 893416
Conjugate DyLight 488
Dilution Flow Cytometry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias antigen p97 (melanoma associated) identified by monoclonal antibodies 133.2 and96.5, CD228, CD228 antigen, FLJ38863, MAP97Melanoma-associated antigen p97, melanotransferrin, membrane-bound transferrin-like protein, MGC4856, MTF1
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human Melanotransferrin/CD228, Gly20-Gly711, Accession # P08582
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4241
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.