Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melatonin Receptor 1B Antibody, DyLight 550, Novus Biologicals™
SDP

Catalog No. NLS932R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NLS932R 0.05 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NLS932R Supplier Novus Biologicals Supplier No. NLS932R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Melatonin Receptor 1B Polyclonal specifically detects Melatonin Receptor 1B in Human, Mouse, Rat, Chicken samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.

Specifications

Antigen Melatonin Receptor 1B
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), KnockDown
Classification Polyclonal
Conjugate DyLight 550
Dilution Western Blot 1:1000, Immunohistochemistry 1:100, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen, Knockdown Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P49286
Gene Alias FGQTL2, Mel 1b melatonin receptor, Mel1b receptor, MEL-1B-R, melatonin receptor 1B, melatonin receptor MEL1B, melatonin receptor type 1B, MT2
Gene Symbols MTNR1B
Host Species Rabbit
Immunogen A synthetic peptide made to an internal region of the human Melatonin Receptor 1B protein (within residues 200-300) [Swiss-Prot P49286].
Purification Method Affinity Purified
Quantity 0.05 mL
Regulatory Status RUO
Research Discipline GPCR, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 4544
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.