Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melphalan-modified DNA Antibody (MP5/73), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP250504C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP250504C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP250504C Supplier Novus Biologicals Supplier No. NBP250504C
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Melphalan-modified DNA Monoclonal specifically detects Melphalan-modified DNA in Non-species specific samples. It is validated for ELISA, Immunohistochemistry.

Specifications

Antigen Melphalan
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone MP5/73
Conjugate DyLight 650
Dilution ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Rat
Immunogen Native DNA reacted with melphalan
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Target Species Other
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.