Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Melphalan-modified DNA Antibody (MP5/73), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB309332 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB309332 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB309332 Supplier Novus Biologicals Supplier No. NBP250504MFV450
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Melphalan-modified DNA Monoclonal antibody specifically detects Melphalan-modified DNA in Nonspecies specific samples. It is validated for ELISA, Immunohistochemistry

Specifications

Antigen Melphalan-modified DNA
Applications ELISA, Immunohistochemistry
Classification Monoclonal
Clone MP5/73
Conjugate mFluor Violet 450 SE
Dilution ELISA, Immunohistochemistry
Formulation 50mM Sodium Borate
Host Species Rat
Immunogen Native DNA reacted with melphalan
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.