Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Memantine hydrochloride

Catalog No. aaj63830mf
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ63830MF 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ63830MF Supplier Thermo Scientific Chemicals Supplier No. J63830MF
Only null left
Add to Cart

CAS: 41100-52-1 | C12H22ClN | 215.77 g/mol

An NMDA receptor antagonist; stimulates dopamine release

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41100-52-1
Molecular Formula C12H22ClN
Molecular Weight (g/mol) 215.77
MDL Number MFCD00214336
InChI Key LDDHMLJTFXJGPI-UHFFFAOYNA-N
Synonym memantine hydrochloride, memantine hcl, 3,5-dimethyladamantan-1-amine hydrochloride, 3,5-dimethyl-1-adamantanamine hydrochloride, namenda, akatinol, axura, namenda xr, memantine.hcl
PubChem CID 181458
ChEBI CHEBI:64323
IUPAC Name hydrogen 3,5-dimethyladamantan-1-amine chloride
SMILES [H+].[Cl-].CC12CC3CC(C)(C1)CC(N)(C3)C2

Specifications

Color White
Quantity 50 mg
Merck Index 14,5829
Formula Weight 215.76
Physical Form Powder
Chemical Name or Material Memantine hydrochloride
Hazard Category H315-H319
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 255-219-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.