Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bulwark™ Men's Lightweight Khaki Flame Resistant Cargo Pant

Catalog No. 17103762 Shop All Red Kap Industries Products
Click to view available options
Size:
40 x 37 in.

Catalog No. 17-103-762

Supplier: Bulwark™ PMU2KH 40 37U

Only null left
Add to Cart
Add to Cart

Men's Lightweight Khaki Flame Resistant Cargo Pant has six total pockets, including a cargo pocket on each leg to ensure you have plenty of room to store all your tools.

  • Ideal for storage and breathability
  • Sits at the waist
  • Regular fit in thigh and seat with slightly tapered leg
  • Two front slack-style pockets and two cargo utility pockets
  • Left cargo pocket has two snap closures and right utility pocket is open
  • Hazard: Flash Fire, Arc Flash
  • Waistband with hook and eye closure
  • Wash Care: Industrial Laundry - Light Soil, Home Wash
  • Fabric: Flame resistant, 7 oz. (235 g/m2)
  • Blend: 48% Modacrylic/37% Lyocell/15% Aramid
  • Product Collection: Cooltouch 2
  • Country of Origin: Imported, Made in USA Fabric

Specifications

Size 40 x 37 in.
Gender Men's
Material Modacrylic, Lyocell, Aramid
Color Khaki
Seam Type Stitched
No. of Pockets 6
Closure Type Hook and Eye
Disposable Reusable
Hazard Risk Category Category 2
Arc Flash Rating 9
Certifications/Compliance NFPA 2112
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.