Learn More
Bulwark™ Men's Midweight Excel FR™ ComforTouch™ Deluxe Insulated Bib Overall
Shop All Red Kap Industries ProductsDescription
- For cold jobs, this midweight insulated bib overall offers unbeatable warmth with CAT 3 AR/FR protection and deluxe features like rugged, double-reinforced knees.
- Large patch pockets and bonus utility pockets ensure there's a place for all of your tools.
- Certifications Level 1: NFPA 2112, NFPA 70E
- Certifications Level 2: Category 3 Protection, NFPA 2112 - 2018 Compliant
- Hazard: Flash Fire, Arc Flash
- Primary Closure: Concealed Zipper
- Wash Care: Industrial Laundry - Light Soil, Home Wash
Fabric: Shell: Flame resistant, 7 oz. (235 g/m&superscript_2;) Twill, Water-repellent, Insulation Package: 14 oz. (474 g/m&superscript_2;) - Blend: Shell: 88% Cotton/ 12% Nylon, Insulation Package: 50% Modacrylic 50% Cotton
- Closure: Concealed Nomex™ taped brass zipper front
- Pocket: Two large front bib pockets with pencil stall, Two large hip pockets, One rule pocket on right leg, Two front pockets
- Features: Reinforced double knees, Elasticized shoulder straps with non-conductive adjustable hardware, Two large center front pockets with pencil stall
- Product Collection: Excel FR Comfortouch
- Protection: ATPV Arc Rating: 36
- Country of Origin: Imported, Made in USA Fabric
- Fit Details: Designed to be worn over other clothing. Fit over work boots.
- Gender: Male
- Materials: Cotton/Nylon
- Brand: Bulwark™
Specifications
Specifications
| Closure Type | Zipper |
| Color | Gray |
| Material | Cotton, Nylon |
| Seam Type | Stitched |
| Size | Large |
| Suspender | Adjustable |
| Arc Flash Rating | 36 |
| No. of Pockets | 7 |
| Certifications/Compliance | NFPA 2112, NFPA 70E |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.