Learn More
Description
- Three-button front placket for enhanced appearance
- Contrasting pattern on sleeve for athletic look
- CAT 2/2112 left sleeve label leaves no question that you are protected on the job site
- Hazard: Flash Fire, Arc Flash
- Wash Care: Home Wash
- Fabric: Body: 6.75 oz. (230 g/m2) Flame Resistant Jersey, Contrast Panels: 6.75 oz. (230 g/m2) Flame Resistant Jersey
- Blend: Body: 93% Cotton/7% Spandex Elastane, Contrast Panels: 93% Cotton/7% Spandex Elastane
- Collar: Rib
- Country of Origin: Imported, Made in USA Fabric
Specifications
Specifications
| Size | 2X-Large |
| Gender | Men's |
| Material | Cotton, Spandex |
| Color | Navy |
| Neck Style | Henley |
| Sleeve Style | Long Sleeve |
| Construction | Knit |
| Closure Type | Button |
| Disposable | Reusable |
| No. of Pockets | 1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.